Please login to your online account to display your discounted pricing

1-[3-(4-chlorophenyl)-5-methylisoxazol-4-yl]ethan-1-one, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB04957DA

 View more versions of this product

Catalog No. SPB04957DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 235.67
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 1-[3-(4-Chlorophenyl)-5-methylisoxazol-4-yl]ethan-1-one
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C12H10ClNO2
MDL Number MFCD01313507
CAS 169814-48-6

Specifications

Chemical Name or Material 1-[3-(4-Chlorophenyl)-5-methylisoxazol-4-yl]ethan-1-one
CAS 169814-48-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C12H10ClNO2
Formula Weight 235.67
MDL Number MFCD01313507