missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,4,6-Tetrakis(methoxymethyl)glycoluril, TCI America™

Manufacturer:  TCI America T205825G

 View more versions of this product

Catalog No. T205825G



Specifications

Specifications

1,3,4,6-Tetrakis(methoxymethyl)glycoluril
118°C
17464-88-9
MFCD00402438
COCN1C2C(N(C1=O)COC)N(C(=O)N2COC)COC
318.33
318.33
≥98.0% (N)
White
25g
C12H22N4O6
XGQJGMGAMHFMAO-UHFFFAOYSA-N
1,3,4,6-tetrakis(methoxymethyl)-3a,6a-dihydroimidazo[4,5-d]imidazole-2,5-dione
87125
Crystal-Powder at 20°C

Chemical Identifiers

17464-88-9
318.33
XGQJGMGAMHFMAO-UHFFFAOYSA-N
1,3,4,6-tetrakis(methoxymethyl)-3a,6a-dihydroimidazo[4,5-d]imidazole-2,5-dione
C12H22N4O6
MFCD00402438
87125
COCN1C2C(N(C1=O)COC)N(C(=O)N2COC)COC
Safety and Handling

Safety and Handling

TSCA : Yes

Documents