Please login to your online account to display your discounted pricing

1-(3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)piperidine, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC34039DA

 View more versions of this product

Catalog No. CC34039DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 287.21
Quantity 1g
Assay 97%
Molecular Formula C17H26BNO2
CAS 852227-97-5
Chemical Name or Material 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
Packaging Amber Glass Bottle
Assay Percent Range 95%

Specifications

Chemical Name or Material 1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperidine
Assay 97%
Assay Percent Range 95%
Molecular Formula C17H26BNO2
Formula Weight 287.21