Learn More
1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-pyrazole, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC32939CB
Specifications
1-[3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-1H-pyrazole | |
97% | |
C15H19BN2O2 | |
1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 3-pyrazol-1-yl-phenylboronic acid pinacol ester, 3-1h-pyrazol-1-yl benzeneboronic acid, pinacol ester, 3-1h-pyrazol-1-yl phenyl boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-3-pyrazolylphenyl-1,3,2-dioxaborolane, 1-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl-1h-pyrazole, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaboranlan-2-yl phenyl-1h-pyrazole | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N3C=CC=N3 | |
270.139 | |
270.14 |
852227-94-2 | |
Amber Glass Bottle | |
250mg | |
GVSCNAOZDQCWJJ-UHFFFAOYSA-N | |
1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrazole | |
4961249 | |
97% |
Chemical Identifiers
852227-94-2 | |
270.139 | |
1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1h-pyrazole, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 1-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl pyrazole, 3-pyrazol-1-yl-phenylboronic acid pinacol ester, 3-1h-pyrazol-1-yl benzeneboronic acid, pinacol ester, 3-1h-pyrazol-1-yl phenyl boronic acid pinacol ester, 4,4,5,5-tetramethyl-2-3-pyrazolylphenyl-1,3,2-dioxaborolane, 1-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl-1h-pyrazole, 1-3-4,4,5,5-tetramethyl-1,3,2-dioxaboranlan-2-yl phenyl-1h-pyrazole | |
1-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pyrazole |
C15H19BN2O2 | |
GVSCNAOZDQCWJJ-UHFFFAOYSA-N | |
4961249 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)N3C=CC=N3 |
Safety and Handling
GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning