Please login to your online account to display your discounted pricing

1,3,3-Trimethyl-2-methyleneindoline 98.5%, ACROS Organics™


Quantity 250g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  421942500

 View more versions of this product

Catalog No. AC421942500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 173.26
Quantity 250g
Color Clear red
EINECS Number 204-235-1
TSCA TSCA
RTECS Number NM1926500
Density 0.986g/mL
CAS Min % 98.0
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.5760 to 1.5780 (20°C, 589nm)
Synonym Fischers base
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material 1, 3, 3-Trimethyl-2-methyleneindoline
Name Note 98.5%
Assay Percent Range 98.50%
Molecular Formula C12H15N
MDL Number MFCD00005813
CAS 118-12-7

Specifications

Chemical Name or Material 1, 3, 3-Trimethyl-2-methyleneindoline
Name Note 98.5%
CAS 118-12-7
CAS Min % 98.0
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98.50%
Molecular Formula C12H15N
Formula Weight 173.26
MDL Number MFCD00005813
Synonym Fischers base
Accessories

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents