Please login to your online account to display your discounted pricing

1,3,3-Trimethyl-2-methyleneindoline 98.5%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  421941000

 View more versions of this product

Catalog No. AC421941000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 173.26
Quantity 100g
Color Clear red
TSCA TSCA
RTECS Number NM1926500
EINECS Number 204-235-1
Density 0.986g/mL
CAS Min % 98.0
CAS Max % 100.0
Synonym Fischers base
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Refractive Index 1.5760 to 1.5780 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
CAS 118-12-7
Name Note 98.5%
Chemical Name or Material 1, 3, 3-Trimethyl-2-methyleneindoline
MDL Number MFCD00005813
Molecular Formula C12H15N
Assay Percent Range 98.50%

Specifications

Chemical Name or Material 1, 3, 3-Trimethyl-2-methyleneindoline
Name Note 98.5%
CAS 118-12-7
CAS Min % 98.0
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98.50%
Molecular Formula C12H15N
Formula Weight 173.26
MDL Number MFCD00005813
Synonym Fischers base
Accessories

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.

SDS & Safety Documents