missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,3,3-Trimethyl-2-methyleneindoline 98.5%, ACROS Organics™

$81.35 - $81.35

Chemical Identifiers

CAS 118-12-7
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98.50%
Molecular Formula C12H15N
Formula Weight 173.26
MDL Number MFCD00005813
Synonym Fischers base
Chemical Name or Material 1, 3, 3-Trimethyl-2-methyleneindoline, 98.5%
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC421941000 SDS Thermo Fisher Scientific
421941000
Glass bottle 100g Each for $81.35
AC421942500 SDS Thermo Fisher Scientific
421942500
Glass bottle 250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

118-12-7
100.0
C12H15N
MFCD00005813
1, 3, 3-Trimethyl-2-methyleneindoline, 98.5%
98.0
98.50%
173.26
Fischers base
≥97.5%

Specifications

0.986g/mL
100g
98.0
Authentic
1.5760 to 1.5780 (20°C, 589nm)
98.50%
173.26
Fischers base
≥97.5%
Clear red
118-12-7
100.0
Glass bottle
Liquid
C12H15N
MFCD00005813
1, 3, 3-Trimethyl-2-methyleneindoline, 98.5%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 204-235-1

RTECSNumber : NM1926500

TSCA : TSCA