missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1(2H)-Phthalazinone, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC384575000

 View more versions of this product

Catalog No. AC384575000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

1(2H)-Phthalazinone
184° to 187°C
119-39-1
100.0
Glass bottle
≥98.0%
C8H6N2O
MFCD00006892
Off-white to pale yellow
500g
98.0
Authentic
Crystalline Powder
98+%
146.15

Chemical Identifiers

119-39-1
100.0
C8H6N2O
MFCD00006892
≥98.0%
98.0
98+%
146.15
1(2H)-Phthalazinone

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
Suspected of causing genetic defects.
Causes skin irritation.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.