Please login to your online account to display your discounted pricing

1(2H)-Phthalazinone, 98+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC384570250

 View more versions of this product

Catalog No. AC384570250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Crystalline Powder
Formula Weight 146.15
Quantity 25g
Color Off-white to pale yellow
Melting Point 184° to 187°C
CAS Min % 98.0
CAS Max % 100.0
Chemical Name or Material 1(2H)-Phthalazinone
Assay Percent Range 98+%
Molecular Formula C8H6N2O
CAS 119-39-1
MDL Number MFCD00006892
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Suspected of causing genetic defects.
Causes skin irritation.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 1(2H)-Phthalazinone
CAS 119-39-1
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C8H6N2O
Formula Weight 146.15
MDL Number MFCD00006892
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Suspected of causing genetic defects.
Causes skin irritation.
Harmful if swallowed.

SDS & Safety Documents