Learn More
1(2H)-Phthalazinone, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 384570050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1(2H)-Phthalazinone | |
98.0 | |
184.0°C to 187.0°C | |
Authentic | |
Glass bottle | |
MFCD00006892 | |
1 2h-phthalazinone, phthalazin-1 2h-one, phthalazone, phthalazinone, phthalazin-1-one, 1-phthalazinol, phthalazin-1-ol, 1-hydroxyphthalazine, 1 2h phthalazinone, 1,2-dihydrophthalazin-1-one | |
C1=CC=C2C(=C1)C=NNC2=O | |
146.15 | |
CHEBI:34023 | |
98+% |
119-39-1 | |
100.0 | |
White to Yellow | |
98% min. (HPLC) | |
C8H6N2O | |
5 g | |
IJAPPYDYQCXOEF-UHFFFAOYSA-N | |
2H-phthalazin-1-one | |
8394 | |
146.15 | |
Crystalline Powder |
Chemical Identifiers
119-39-1 | |
146.15 | |
IJAPPYDYQCXOEF-UHFFFAOYSA-N | |
8394 | |
2H-phthalazin-1-one |
C8H6N2O | |
MFCD00006892 | |
1 2h-phthalazinone, phthalazin-1 2h-one, phthalazone, phthalazinone, phthalazin-1-one, 1-phthalazinol, phthalazin-1-ol, 1-hydroxyphthalazine, 1 2h phthalazinone, 1,2-dihydrophthalazin-1-one | |
CHEBI:34023 | |
C1=CC=C2C(=C1)C=NNC2=O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Warning
EINECSNumber : 204-319-8
RUO – Research Use Only