missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Thienyl)ethylamine, tech. 90%

Chemical Identifiers

CAS 6309-16-6
Molecular Formula C6H9NS
Formula Weight 127.21
MDL Number MFCD02734311
UN Number UN2735
Synonym alpha-Methyl-2-thiophenemethylamine
Chemical Name or Material 1-(2-Thienyl)ethylamine
Grade Technical Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH26052ME SDS Alfa Aesar™
H26052ME
500mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

6309-16-6
127.21
UN2735
1-(2-Thienyl)ethylamine
C6H9NS
MFCD02734311
alpha-Methyl-2-thiophenemethylamine
Technical Grade

Specifications

500mg
C6H9NS
MFCD02734311
alpha-Methyl-2-thiophenemethylamine
90%
6309-16-6
127.21
UN2735
1-(2-Thienyl)ethylamine
Technical Grade

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.