missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Tetrahydropyranyl)-3-(trifluoromethyl)-1H-pyrazole-5-boronic acid, 98%

Manufacturer:  Alfa Aesar™ H64012MD

 View more versions of this product

Catalog No. AAH64012MD



Specifications

Specifications

250mg
C9H12BF3N2O3
1-oxan-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl boronic acid, 1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl boronic acid, 1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl boronicacid, 1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-ylboronic acid, 1-tetrahydropyran-2-yl-3-trifluoromethyl pyrazole-5-boronic acid, 2-oxan-2-yl-5-trifluoromethyl pyrazol-3-ylboronic acid, 2-tetrahydropyran-2-yl-5-trifluoromethyl pyrazol-3-yl boronic acid, acmc-2099kl, boronic acid, b-1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl
B(C1=CC(=NN1C2CCCCO2)C(F)(F)F)(O)O
264.011
264.01
98%
1141878-45-6
MFCD17214248
IIRVABFYRULQMI-UHFFFAOYSA-N
[2-(oxan-2-yl)-5-(trifluoromethyl)pyrazol-3-yl]boronic acid
53216481
Solid

Chemical Identifiers

1141878-45-6
264.011
IIRVABFYRULQMI-UHFFFAOYSA-N
53216481
B(C1=CC(=NN1C2CCCCO2)C(F)(F)F)(O)O
C9H12BF3N2O3
MFCD17214248
1-oxan-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl boronic acid, 1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl boronic acid, 1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl boronicacid, 1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-ylboronic acid, 1-tetrahydropyran-2-yl-3-trifluoromethyl pyrazole-5-boronic acid, 2-oxan-2-yl-5-trifluoromethyl pyrazol-3-ylboronic acid, 2-tetrahydropyran-2-yl-5-trifluoromethyl pyrazol-3-yl boronic acid, acmc-2099kl, boronic acid, b-1-tetrahydro-2h-pyran-2-yl-3-trifluoromethyl-1h-pyrazol-5-yl
[2-(oxan-2-yl)-5-(trifluoromethyl)pyrazol-3-yl]boronic acid
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

Documents