missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Tetrahydropyranyl)-1H-pyrazole, 98%

Manufacturer:  Alfa Aesar™ H6424606

 View more versions of this product

Catalog No. AAH6424606



Specifications

Specifications

1-(2-Tetrahydropyranyl)-1H-pyrazole
1.5030
Colorless to Yellow
449758-17-2
MFCD11044692
1-2-tetrahydropyranyl-1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl-1h-pyrazole, 1-oxan-2-yl-1h-pyrazole, 1-oxan-2-yl pyrazole, 1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl, 1-2-tetrahydropyranyl pyrazole, 1-tetrahydro-pyran-2-yl-1h-pyrazole, 1-2-oxanyl pyrazole, ksc588k5l, 1-tetrahydropyran-2-ylpyrazole
C1CCOC(C1)N2C=CC=N2
152.197
152.2
98%
1.084g/mL
60°C to 65°C (0.8mmHg)
5g
C8H12N2O
9694988
IMZWSOSYNFVECD-UHFFFAOYSA-N
1-(oxan-2-yl)pyrazole
21866310
Liquid

Chemical Identifiers

449758-17-2
152.197
IMZWSOSYNFVECD-UHFFFAOYSA-N
21866310
C1CCOC(C1)N2C=CC=N2
C8H12N2O
MFCD11044692
1-2-tetrahydropyranyl-1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl-1h-pyrazole, 1-oxan-2-yl-1h-pyrazole, 1-oxan-2-yl pyrazole, 1h-pyrazole, 1-tetrahydro-2h-pyran-2-yl, 1-2-tetrahydropyranyl pyrazole, 1-tetrahydro-pyran-2-yl-1h-pyrazole, 1-2-oxanyl pyrazole, ksc588k5l, 1-tetrahydropyran-2-ylpyrazole
1-(oxan-2-yl)pyrazole
SDS Documents