Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid neopentyl glycol ester, 95%

$171.77 - $390.26

Chemical Identifiers

Chemical Name or Material 1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid neopentyl glycol ester
CAS 1072944-26-3
Molecular Formula C13H21BN2O3
Formula Weight 264.13
MDL Number MFCD09037503
Synonym 4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-(tetrahydropyran-2-yl)-1H-pyrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2778303

  SDS
alfa aesar™
H2778303
1g Each for $390.26

AAH27783MD

  SDS
alfa aesar™
H27783MD
250mg Each for $171.77
Description & Specifications

Specifications

Formula Weight 264.13
Chemical Name or Material 1-(2-Tetrahydropyranyl)-1H-pyrazole-4-boronic acid neopentyl glycol ester
Melting Point 84° to 91°C
CAS 1072944-26-3
Synonym 4-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-(tetrahydropyran-2-yl)-1H-pyrazole
Quantity 1g
Assay 95%
MDL Number MFCD09037503
Molecular Formula C13H21BN2O3