Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Tetrahydrofuroyl)piperazine, 97%

$54.03 - $166.24

Chemical Identifiers

Chemical Name or Material 1-(2-Tetrahydrofuroyl)piperazine
CAS 63074-07-7
Molecular Formula C9H16N2O2
Formula Weight 184.24
MDL Number MFCD00190247
Beilstein 880578
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1870406

 
alfa aesar™
L1870406
5g Each for $54.03

AAL1870414

 
alfa aesar™
L1870414
25g Each for $166.24
Description & Specifications

Specifications

Formula Weight 184.24
CAS 63074-07-7
Chemical Name or Material 1-(2-Tetrahydrofuroyl)piperazine
Quantity 5g
Assay 97%
MDL Number MFCD00190247
Health Hazard 1 Warning
Molecular Formula C9H16N2O2
Beilstein 880578
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 120° to 125°C (0.2mmHg)

Applications
An intermediate of terazosin hydrochloride. It has also been used as a reactant for the preparation of pyrazol-3-propanoic acid derivatives as inhibitors of leukotriene biosynthesis in human neutrophils.

Solubility
Fully miscible in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.