Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Pyrimidinyl)piperidine-4-carboxylic acid, 95%

$105.03 - $361.62

Chemical Identifiers

Chemical Name or Material 1-(2-Pyrimidinyl)piperidine-4-carboxylic acid
CAS 303144-44-7
Molecular Formula C10H13N3O2
Formula Weight 207.23
MDL Number MFCD00664474
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5035103

  SDS
alfa aesar™
H5035103
1g Each for $361.62

AAH50351MD

  SDS
alfa aesar™
H50351MD
250mg Each for $105.03
Description & Specifications

Specifications

Formula Weight 207.23
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
CAS 303144-44-7
Quantity 1g
Molecular Formula C10H13N3O2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 1-(2-Pyrimidinyl)piperidine-4-carboxylic acid
Assay 95%
MDL Number MFCD00664474
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.