Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Pyrimidinyl)piperazine dihydrochloride, 98+%

$28.18 - $116.32

Chemical Identifiers

Chemical Name or Material 1-(2-Pyrimidinyl)piperazine dihydrochloride
CAS 94021-22-4
Molecular Formula C8H12N4·2HCl
Formula Weight 237.13
MDL Number MFCD00012779
Beilstein 151178
Synonym 2-(1-Piperazinyl)pyrimidine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2408506

  SDS
alfa aesar™
B2408506
5g Each for $28.18

AAB2408514

  SDS
alfa aesar™
B2408514
25g Each for $116.32
Description & Specifications

Specifications

Formula Weight 237.13
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00012779
Quantity 5g
Molecular Formula C8H12N4·2HCl
Melting Point ≈280°C (dec.)
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
RTECS Number UV9702000
Synonym 2-(1-Piperazinyl)pyrimidine dihydrochloride
Beilstein 151178
Chemical Name or Material 1-(2-Pyrimidinyl)piperazine dihydrochloride
Assay ≥98%
CAS 94021-22-4
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.