missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Pyrimidinyl)homopiperazine

Chemical Identifiers

CAS 21279-57-2
Molecular Formula C9H14N4
Formula Weight 178.24
MDL Number MFCD06738258
UN Number UN2735
Synonym 1-(2-Pyrimidinyl)-1,4-diazepane
Chemical Name or Material 1-(2-Pyrimidinyl)homopiperazine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5016103 SDS Alfa Aesar™
H5016103
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH50161MD SDS Alfa Aesar™
H50161MD
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

21279-57-2
178.24
UN2735
1-(2-Pyrimidinyl)homopiperazine
C9H14N4
MFCD06738258
1-(2-Pyrimidinyl)-1,4-diazepane

Specifications

1g
C9H14N4
MFCD06738258
1-(2-Pyrimidinyl)-1,4-diazepane
21279-57-2
178.24
UN2735
1-(2-Pyrimidinyl)homopiperazine

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.