Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Pyridyl)ethylamine, 96%

Chemical Identifiers

Chemical Name or Material 1-(2-Pyridyl)ethylamine
CAS 42088-91-5
Molecular Formula C7H10N2
Formula Weight 122.17
MDL Number MFCD05215238
UN Number UN2735
Synonym alpha-Methyl-2-pyridinemethylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2604603

 
alfa aesar™
H2604603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH26046MD

 
alfa aesar™
H26046MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.17
Chemical Name or Material 1-(2-Pyridyl)ethylamine
Assay 96%
Molecular Formula C7H10N2
MDL Number MFCD05215238
CAS 42088-91-5
UN Number UN2735
Synonym alpha-Methyl-2-pyridinemethylamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.