missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Propanediol OR, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 57-55-6
Molecular Formula CH3CH(OH)CH2OH
Formula Weight 76.1
Chemical Name or Material 1, 2-Propanediol
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK19251 Avantor Performance
192561
Glass 1kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

57-55-6
CH3CH(OH)CH2OH
76.1
1, 2-Propanediol

Specifications

76.1
Glass
1kg
57-55-6
CH3CH(OH)CH2OH
1, 2-Propanediol