Please login to your online account to display your discounted pricing

1,2-Propanediol, EMD Millipore

For General Chemistry

$908.91 - $1,707.09

Chemical Identifiers

Chemical Name or Material 1,2-Propanediol
CAS 57-55-6
Physical Form Liquid
Assay ≥98.0%
Molecular Formula CH3H8O2
Formula Weight 76.1
Synonym Propylene Glycol
Color Clear
Melting Point −60°C
Solubility Information Water: Soluble
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MPX16903

 
emd millipore corporation
PX16903
Glass Bottle 4L Each for $908.91

MPX16905

 
emd millipore corporation
PX16905
Poly Pail 18.9L Each for $1,707.09
Description & Specifications

Specifications

Melting Point −60°C
Assay ≥98.0%
Boiling Point 187.4°C
Formula Weight 76.1
Quantity 4L
Color Clear
Recommended Storage Room Temperature. Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.
CAS 57-55-6
Synonym Propylene Glycol
Flash Point 98.85°C (209.9°F)
Molecular Formula CH3H8O2
Packaging Glass Bottle
Physical Form Liquid
Solubility Information Water: Soluble
Chemical Name or Material 1,2-Propanediol
Safety and Handling

Room Temperature. Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.