Please login to your online account to display your discounted pricing

1,2-Propanediol, ACS, 99.5%, Alfa Aesar

As a substitute for ethylene glycol and glycerol

$51.72 - $3,326.09

Chemical Identifiers

Chemical Name or Material 1, 2-Propanediol
CAS 57-55-6
Purity Grade ACS Grade
Physical Form Liquid
Assay Percent Range 99.5%
Molecular Formula CH3CH(OH)CH2OH
Formula Weight 76.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA309481Q

 
alfa aesar
309481Q
55 gal. Each for $3,326.09

AA30948AP

 
alfa aesar
30948AP
500mL Each for $51.72
Description & Specifications

Specifications

Formula Weight 76.1
Quantity 55 gal.
Purity Grade ACS Grade
Density 1.036g/mL
Melting Point −60°C (−76°F)
Boiling Point 186 to 188°
Flash Point 107°C
Chemical Name or Material 1, 2-Propanediol
Physical Form Liquid
Assay Percent Range 99.5%
Molecular Formula CH3CH(OH)CH2OH
CAS 57-55-6
Storage Note 1 Hygroscopic
Safety and Handling

Hygroscopic