Learn More
1,2-Phthalic dicarboxaldehyde, 98+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 131085000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
1, 2-Phthalic dicarboxaldehyde | |
98.0 | |
83°C to 84°C (1.0 hPa) | |
Authentic | |
Glass bottle | |
C6H4(CHO)2 | |
500g | |
15, 7480 | |
Solubility in water: soluble. | |
O=CC1=CC=CC=C1C=O | |
134.13 | |
2.3 mPa.s (65°C) | |
134.13 |
643-79-8 | |
100.0 | |
132°C | |
98% min. (GC) | |
C8H6O2 | |
MFCD00003335 | |
07, 674 | |
o-phthalaldehyde, o-phthaldialdehyde, benzene-1,2-dicarboxaldehyde, 1,2-benzenedicarboxaldehyde, phthaldialdehyde, phthalic aldehyde, phthalic dialdehyde, phthalyldicarboxaldehyde, ortho-phthalaldehyde, o-phthaldehyde | |
ZWLUXSQADUDCSB-UHFFFAOYSA-N | |
phthalaldehyde | |
4807 | |
CHEBI:70851 | |
98+% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
EINECSNumber : 211-402-2
RUO â Research Use Only