Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Phenylethyl)-4-(phenylethynyl)benzene, 97%

$340.53 - $340.53

Chemical Identifiers

Chemical Name or Material 1-(2-Phenylethyl)-4-(phenylethynyl)benzene
CAS 906650-60-0
Molecular Formula C22H18
Formula Weight 282.39
MDL Number MFCD12407075
Synonym 4-Phenethyldiphenylacetylene; 4-Phenylethynylbibenzyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3005803

  SDS
alfa aesar™
H3005803
1g Each for $340.53
Description & Specifications

Specifications

Formula Weight 282.39
Chemical Name or Material 1-(2-Phenylethyl)-4-(phenylethynyl)benzene
Assay 97%
Molecular Formula C22H18
MDL Number MFCD12407075
CAS 906650-60-0
Synonym 4-Phenethyldiphenylacetylene; 4-Phenylethynylbibenzyl
Melting Point 104° to 106°C
Quantity 1g