missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Phenylenediacetic acid, 99%, Acros Organics

Manufacturer:  Thermo Fisher Scientific 157550050

 View more versions of this product

Catalog No. AC157550050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

1, 2-Phenylenediacetic acid
Glass Bottle
150°C to 152°C
99%
98.5
99%
C6H4(CH2CO2H)2
09, 874
MMEDJBFVJUFIDD-UHFFFAOYSA-N
2-[2-(carboxymethyl)phenyl]acetic acid
24123
Needle-Like Crystalline Powder
Authentic
Beige to Beige-Brown
5g
7500-53-0
100.0
C10H10O4
MFCD00004329
1,2-benzenediacetic acid
C1=CC=C(C(=C1)CC(=O)O)CC(=O)O
194.186
194.19
99%

Chemical Identifiers

7500-53-0
194.186
MMEDJBFVJUFIDD-UHFFFAOYSA-N
24123
C1=CC=C(C(=C1)CC(=O)O)CC(=O)O
C10H10O4
MFCD00004329
1,2-benzenediacetic acid
2-[2-(carboxymethyl)phenyl]acetic acid
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 231-355-1

TSCA : TSCA

SDS Documents