Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Phenylene phosphorotrichloridite, 94%

Chemical Identifiers

Chemical Name or Material 1,2-Phenylene phosphorotrichloridite
CAS 2007-97-8
Molecular Formula C6H4Cl3O2P
Formula Weight 245.43
MDL Number MFCD00014578
UN Number UN3261
Beilstein 743959
Synonym Catechyl phosphorus trichloride; 2,2,2-Trichloro-1,3,2-benzodioxaphosphole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0022714

  SDS
alfa aesar™
L0022714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 245.43
Chemical Name or Material 1,2-Phenylene phosphorotrichloridite
MDL Number MFCD00014578
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 94%
Boiling Point 102° to 104°C (2mmHg)
Molecular Formula C6H4Cl3O2P
UN Number UN3261
Beilstein 743959
CAS 2007-97-8
Synonym Catechyl phosphorus trichloride; 2,2,2-Trichloro-1,3,2-benzodioxaphosphole
Odor Acrid
Health Hazard 1 Danger
Melting Point 48° to 50°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Quantity 25g
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.