missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1-(2-Phenyl-1,3-thiazol-4-yl)ethan-1-one, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals SPB03431EA
Specifications
1-(2-Phenyl-1, 3-thiazol-4-yl)ethan-1-one | |
95.0 | |
C11H9NOS | |
10g | |
ZOOGZFPRAKXWKI-UHFFFAOYSA-N | |
1-(2-phenyl-1,3-thiazol-4-yl)ethanone | |
604819 | |
95% |
10045-52-0 | |
95% | |
MFCD00068112 | |
1-2-phenylthiazol-4-yl ethanone, 1-2-phenyl-1,3-thiazol-4-yl ethan-1-one, 1-2-phenyl-1,3-thiazol-4-yl ethanone, 4-acetyl-2-phenylthiazole, 1-2-phenyl-1,3-thiazol-4-yl-1-ethanone, ethanone, 1-2-phenyl-4-thiazolyl, 4-acetyl-2-phenyl-1,3-thiazole, maybridge1_008631, buttpark 15357-18, 2-phenyl-4-acetylthiazole | |
CC(=O)C1=CSC(=N1)C2=CC=CC=C2 | |
203.259 | |
203.26 |
Chemical Identifiers
10045-52-0 | |
203.259 | |
ZOOGZFPRAKXWKI-UHFFFAOYSA-N | |
604819 | |
CC(=O)C1=CSC(=N1)C2=CC=CC=C2 |
C11H9NOS | |
MFCD00068112 | |
1-2-phenylthiazol-4-yl ethanone, 1-2-phenyl-1,3-thiazol-4-yl ethan-1-one, 1-2-phenyl-1,3-thiazol-4-yl ethanone, 4-acetyl-2-phenylthiazole, 1-2-phenyl-1,3-thiazol-4-yl-1-ethanone, ethanone, 1-2-phenyl-4-thiazolyl, 4-acetyl-2-phenyl-1,3-thiazole, maybridge1_008631, buttpark 15357-18, 2-phenyl-4-acetylthiazole | |
1-(2-phenyl-1,3-thiazol-4-yl)ethanone |