missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
1,2-Pentanediol, 96%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC123290500
Specifications
1, 2-Pentanediol | |
105°C | |
Glass bottle | |
0.971 | |
Colorless to Yellow | |
97% | |
96.0 | |
95% min. (GC) | |
CH3(CH2)2CH(OH)CH2OH | |
1,2-dihydroxypentane, 1,2-pentanediol, 1,2-pentanediol, 2r, 3-01-00-02191 beilstein handbook reference, acmc-1axdb, acmc-20mbh5, ksc271i3n, wcvrqhfdjllwfe-uhfffaoysa | |
WCVRQHFDJLLWFE-UHFFFAOYSA-N | |
pentane-1,2-diol | |
93000 | |
Liquid |
0.9710g/mL | |
Authentic | |
1.4387 to 1.4407 | |
206.0°C | |
50mL | |
5343-92-0 | |
100.0 | |
C5H12O2 | |
01,II,548 | |
Solubility in water: miscible. | |
CCCC(CO)O | |
104.15 | |
104.15 | |
96% |
Chemical Identifiers
5343-92-0 | |
104.15 | |
1,2-dihydroxypentane, 1,2-pentanediol, 1,2-pentanediol, 2r, 3-01-00-02191 beilstein handbook reference, acmc-1axdb, acmc-20mbh5, ksc271i3n, wcvrqhfdjllwfe-uhfffaoysa | |
pentane-1,2-diol |
C5H12O2 | |
WCVRQHFDJLLWFE-UHFFFAOYSA-N | |
93000 | |
CCCC(CO)O |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
EINECSNumber : 226-285-3
RTECSNumber : SA0455000
TSCA : TSCA