missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-O-Cyclohexylidene-alpha-D-glucofuranose, 97%

$51.04 - $181.97

Chemical Identifiers

CAS 16832-21-6
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.286
MDL Number MFCD00142481
InChI Key KAUCUKASFLXURN-RCZSTQMZSA-N
Synonym r-1-3a'r,5'r,6's,6a'r-6'-hydroxytetrahydrospiro cyclohexane-1,2'-furo 2,3-d 1,3 dioxol-5'-yl ethane-1,2-diol, 1r-1-3'ar,5'r,6's,6'ar-6'-hydroxy-tetrahydrospiro cyclohexane-1,2'-furo 2,3-d 1,3 dioxole-5'-yl ethane-1,2-diol, 1,2-o-cyclohexylidene-a-d-glucofuranose, 1,2-o-cyclohexylidene-alpha-d-glucofuranose, 1r-1-3'ar,5'r,6's,6'ar-6'-hydroxy-tetrahydrospiro cyclohexane-1,2'-furo 2,3-d 1,3 dioxol-5'-yl ethane-1,2-diol, 1,2-o-cyclohexylidene-, a-d-glucofuranose, a-d-glucofuranose,1,2-o-cyclohexylidene, 1-o,2-o-cyclohexylidene-alpha-d-glucofuranose, 1,2-o-cyclohexylidene-alpha-d-glucofuranose, 1,2-o-cyclohexane-1,1-diyl-alpha-d-xylo-hexofuranose
PubChem CID 2724778
IUPAC Name (1R)-1-[(3aR,5R,6S,6aR)-6-hydroxyspiro[3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxole-2,1'-cyclohexane]-5-yl]ethane-1,2-diol
SMILES C1CCC2(CC1)OC3C(C(OC3O2)C(CO)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6676103 View Documents Alfa Aesar™
J6676103
1g Each for $51.04
 
AAJ6676106 View Documents Alfa Aesar™
J6676106
5g Each for $181.97
 
Specifications

Chemical Identifiers

16832-21-6
260.286
KAUCUKASFLXURN-RCZSTQMZSA-N
2724778
C1CCC2(CC1)OC3C(C(OC3O2)C(CO)O)O
C12H20O6
MFCD00142481
r-1-3a'r,5'r,6's,6a'r-6'-hydroxytetrahydrospiro cyclohexane-1,2'-furo 2,3-d 1,3 dioxol-5'-yl ethane-1,2-diol, 1r-1-3'ar,5'r,6's,6'ar-6'-hydroxy-tetrahydrospiro cyclohexane-1,2'-furo 2,3-d 1,3 dioxole-5'-yl ethane-1,2-diol, 1,2-o-cyclohexylidene-a-d-glucofuranose, 1,2-o-cyclohexylidene-alpha-d-glucofuranose, 1r-1-3'ar,5'r,6's,6'ar-6'-hydroxy-tetrahydrospiro cyclohexane-1,2'-furo 2,3-d 1,3 dioxol-5'-yl ethane-1,2-diol, 1,2-o-cyclohexylidene-, a-d-glucofuranose, a-d-glucofuranose,1,2-o-cyclohexylidene, 1-o,2-o-cyclohexylidene-alpha-d-glucofuranose, 1,2-o-cyclohexylidene-alpha-d-glucofuranose, 1,2-o-cyclohexane-1,1-diyl-alpha-d-xylo-hexofuranose
(1R)-1-[(3aR,5R,6S,6aR)-6-hydroxyspiro[3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxole-2,1'-cyclohexane]-5-yl]ethane-1,2-diol

Specifications

White
1g
C12H20O6
r-1-3a'r,5'r,6's,6a'r-6'-hydroxytetrahydrospiro cyclohexane-1,2'-furo 2,3-d 1,3 dioxol-5'-yl ethane-1,2-diol, 1r-1-3'ar,5'r,6's,6'ar-6'-hydroxy-tetrahydrospiro cyclohexane-1,2'-furo 2,3-d 1,3 dioxole-5'-yl ethane-1,2-diol, 1,2-o-cyclohexylidene-a-d-glucofuranose, 1,2-o-cyclohexylidene-alpha-d-glucofuranose, 1r-1-3'ar,5'r,6's,6'ar-6'-hydroxy-tetrahydrospiro cyclohexane-1,2'-furo 2,3-d 1,3 dioxol-5'-yl ethane-1,2-diol, 1,2-o-cyclohexylidene-, a-d-glucofuranose, a-d-glucofuranose,1,2-o-cyclohexylidene, 1-o,2-o-cyclohexylidene-alpha-d-glucofuranose, 1,2-o-cyclohexylidene-alpha-d-glucofuranose, 1,2-o-cyclohexane-1,1-diyl-alpha-d-xylo-hexofuranose
C1CCC2(CC1)OC3C(C(OC3O2)C(CO)O)O
260.286
260.28
97%
163°C to 168°C
16832-21-6
MFCD00142481
KAUCUKASFLXURN-RCZSTQMZSA-N
(1R)-1-[(3aR,5R,6S,6aR)-6-hydroxyspiro[3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxole-2,1'-cyclohexane]-5-yl]ethane-1,2-diol
2724778
Powder
1,2-O-Cyclohexylidene-alpha-D-glucofuranose