missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Nitrophenyl)pyrrolidine, 97%

$28.84 - $337.73

Chemical Identifiers

CAS 40832-79-9
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.218
MDL Number MFCD01604393
InChI Key DHTSBEQPOXRZBB-UHFFFAOYSA-N
Synonym 1-2-nitrophenyl pyrrolidine, 2-nitro-1-pyrrolidinobenzene, 1-2-nitro-phenyl-pyrrolidine, 2-nitrophenyl pyrrolidine, o-pyrrolidin-1-ylnitrobenzene, acmc-1cujb, dhtsbeqpoxrzbb-uhfffaoysa
PubChem CID 12414144
IUPAC Name 1-(2-nitrophenyl)pyrrolidine
SMILES C1CCN(C1)C2=CC=CC=C2[N+](=O)[O-]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63984MD View Documents Alfa Aesar™
H63984MD
250mg Each for $28.84
 
AAH6398403 View Documents Alfa Aesar™
H6398403
1g Each for $85.14
 
AAH6398406 View Documents Alfa Aesar™
H6398406
5g Each for $337.73
 
Specifications

Chemical Identifiers

40832-79-9
192.218
DHTSBEQPOXRZBB-UHFFFAOYSA-N
12414144
C1CCN(C1)C2=CC=CC=C2[N+](=O)[O-]
C10H12N2O2
MFCD01604393
1-2-nitrophenyl pyrrolidine, 2-nitro-1-pyrrolidinobenzene, 1-2-nitro-phenyl-pyrrolidine, 2-nitrophenyl pyrrolidine, o-pyrrolidin-1-ylnitrobenzene, acmc-1cujb, dhtsbeqpoxrzbb-uhfffaoysa
1-(2-nitrophenyl)pyrrolidine

Specifications

250mg
C10H12N2O2
1-2-nitrophenyl pyrrolidine, 2-nitro-1-pyrrolidinobenzene, 1-2-nitro-phenyl-pyrrolidine, 2-nitrophenyl pyrrolidine, o-pyrrolidin-1-ylnitrobenzene, acmc-1cujb, dhtsbeqpoxrzbb-uhfffaoysa
C1CCN(C1)C2=CC=CC=C2[N+](=O)[O-]
192.218
192.21
97%
40832-79-9
MFCD01604393
DHTSBEQPOXRZBB-UHFFFAOYSA-N
1-(2-nitrophenyl)pyrrolidine
12414144
Solid
1-(2-Nitrophenyl)pyrrolidine