Alfa Aesar™ 1-(2-Nitrophenyl)pyrrolidine, 97%
Manufacturer:  Alfa Aesar™ H63984MD
 View more versions of this product
Catalog No. AAH63984MD
Specifications
Specifications
1-(2-Nitrophenyl)pyrrolidine
40832-79-9
MFCD01604393
DHTSBEQPOXRZBB-UHFFFAOYSA-N
1-(2-nitrophenyl)pyrrolidine
12414144
Solid
250mg
C10H12N2O2
1-2-nitrophenyl pyrrolidine, 2-nitro-1-pyrrolidinobenzene, 1-2-nitro-phenyl-pyrrolidine, 2-nitrophenyl pyrrolidine, o-pyrrolidin-1-ylnitrobenzene, acmc-1cujb, dhtsbeqpoxrzbb-uhfffaoysa
C1CCN(C1)C2=CC=CC=C2[N+](=O)[O-]
192.218
192.21
97%
Chemical Identifiers
40832-79-9
192.218
DHTSBEQPOXRZBB-UHFFFAOYSA-N
12414144
C1CCN(C1)C2=CC=CC=C2[N+](=O)[O-]
C10H12N2O2
MFCD01604393
1-2-nitrophenyl pyrrolidine, 2-nitro-1-pyrrolidinobenzene, 1-2-nitro-phenyl-pyrrolidine, 2-nitrophenyl pyrrolidine, o-pyrrolidin-1-ylnitrobenzene, acmc-1cujb, dhtsbeqpoxrzbb-uhfffaoysa
1-(2-nitrophenyl)pyrrolidine
Documents