Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Nitrophenyl)pyrrole, 97%

$41.58 - $138.47

Chemical Identifiers

Chemical Name or Material 1-(2-Nitrophenyl)pyrrole
CAS 33265-60-0
Molecular Formula C10H8N2O2
Formula Weight 188.19
MDL Number MFCD00047062
Synonym 2-(1-Pyrrolyl)nitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1271603

  SDS
alfa aesar™
A1271603
1g Each for $41.58

AAA1271606

  SDS
alfa aesar™
A1271606
5g Each for $138.47
Description & Specifications

Specifications

Formula Weight 188.19
Melting Point 58° to 60°C
Quantity 1g
Molecular Formula C10H8N2O2
CAS 33265-60-0
Chemical Name or Material 1-(2-Nitrophenyl)pyrrole
Assay 97%
MDL Number MFCD00047062
Synonym 2-(1-Pyrrolyl)nitrobenzene
Boiling Point 130°C (0.4mmHg)