Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Nitrophenyl)piperidine, 98%

$65.99 - $723.24

Chemical Identifiers

Chemical Name or Material 1-(2-Nitrophenyl)piperidine
CAS 15822-77-2
Molecular Formula C11H14N2O2
Formula Weight 206.24
MDL Number MFCD00023653
Synonym 1-(1-Piperidinyl)-2-nitrobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2511606

  SDS
alfa aesar™
B2511606
5g Each for $65.99

AAB2511614

  SDS
alfa aesar™
B2511614
25g Each for $233.55

AAB2511622

  SDS
alfa aesar™
B2511622
100g Each for $723.24
Description & Specifications

Specifications

Formula Weight 206.24
Chemical Name or Material 1-(2-Nitrophenyl)piperidine
Assay 98%
Molecular Formula C11H14N2O2
MDL Number MFCD00023653
CAS 15822-77-2
Synonym 1-(1-Piperidinyl)-2-nitrobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 76° to 78°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.