missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Nitrophenyl)piperazine 98.0+%, TCI America™

Manufacturer:  TCI America N093525G

 View more versions of this product

Catalog No. N093525G



Specifications

Specifications

1-(2-Nitrophenyl)piperazine
Undesignated
59084-06-9
MFCD00040728
YJRCDSXLKPERNV-UHFFFAOYSA-N
1-(2-nitrophenyl)piperazine
100949
Clear Liquid at 20°C
126°C
25g
C10H13N3O2
1-2-nitrophenyl piperazine, 1-2-nitro-phenyl-piperazine, 2-nitrophenyl piperazine, 1-2-nitrophenyl piperazine, 1-2-nitrophenyl piperazine, 1-2-nitrophenyl piperazin, nitrophenylpiperazine, acmc-20ahsu, pubchem14574, 4-nitrophenyl piperazine
C1CN(CCN1)C2=CC=CC=C2[N+](=O)[O-]
207.233
207.23
≥98.0% (GC)

Chemical Identifiers

59084-06-9
207.233
YJRCDSXLKPERNV-UHFFFAOYSA-N
100949
C1CN(CCN1)C2=CC=CC=C2[N+](=O)[O-]
C10H13N3O2
MFCD00040728
1-2-nitrophenyl piperazine, 1-2-nitro-phenyl-piperazine, 2-nitrophenyl piperazine, 1-2-nitrophenyl piperazine, 1-2-nitrophenyl piperazine, 1-2-nitrophenyl piperazin, nitrophenylpiperazine, acmc-20ahsu, pubchem14574, 4-nitrophenyl piperazine
1-(2-nitrophenyl)piperazine
Safety and Handling

Safety and Handling

TSCA : Yes

Documents