Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Nitrophenyl)piperazine, 95%

$232.04 - $584.62

Chemical Identifiers

Chemical Name or Material 1-(2-Nitrophenyl)piperazine
CAS 59084-06-9
Molecular Formula C10H13N3O2
Formula Weight 207.23
MDL Number MFCD00040728
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2677303

  SDS
alfa aesar™
H2677303
1g Each for $584.62

AAH26773MD

  SDS
alfa aesar™
H26773MD
250mg Each for $232.04
Description & Specifications

Specifications

Formula Weight 207.23
Chemical Name or Material 1-(2-Nitrophenyl)piperazine
Assay 95%
Molecular Formula C10H13N3O2
MDL Number MFCD00040728
CAS 59084-06-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.