missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Nitrophenyl)ethanol, TCI America™

$160.42 - $481.27

Chemical Identifiers

CAS 3205-25-2
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD02683380
InChI Key DSDBYQDNNWCLHL-UHFFFAOYSA-N
Synonym 2-Nitro-alpha-methylbenzyl Alcohol
PubChem CID 97851
IUPAC Name 1-(2-nitrophenyl)ethanol
SMILES CC(C1=CC=CC=C1[N+](=O)[O-])O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N11321G View Documents TCI America
N11321G
1g Each for $160.42
 
N11325G View Documents TCI America
N11325G
5g Each for $481.27
 
Specifications

Chemical Identifiers

3205-25-2
167.164
DSDBYQDNNWCLHL-UHFFFAOYSA-N
97851
CC(C1=CC=CC=C1[N+](=O)[O-])O
C8H9NO3
MFCD02683380
2-Nitro-alpha-methylbenzyl Alcohol
1-(2-nitrophenyl)ethanol

Specifications

107°C
1g
C8H9NO3
2-Nitro-alpha-methylbenzyl Alcohol
CC(C1=CC=CC=C1[N+](=O)[O-])O
167.164
167.16
≥98.0% (GC)
Undesignated
3205-25-2
MFCD02683380
DSDBYQDNNWCLHL-UHFFFAOYSA-N
1-(2-nitrophenyl)ethanol
97851
Clear Liquid at 20°C
1-(2-Nitrophenyl)ethanol
Safety and Handling

Safety and Handling

TSCA : No