Please login to your online account to display your discounted pricing

1-[2-Nitro-4-(trifluoromethyl)phenyl]piperazine, 97%, Maybridge


Quantity 25g

Manufacturer: fisher scientific company  BTB03244EB

 View more versions of this product

Catalog No. BTB03244EB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 275.23
Quantity 25g
Assay 97%
CAS Max % 100.0
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Chemical Name or Material 1-[2-Nitro-4-(trifluoromethyl)phenyl]piperazine
Assay Percent Range 97%
CAS Min % 96.5
MDL Number MFCD00052613
CAS 58315-38-1
Molecular Formula C11H12F3N3O2

Specifications

Chemical Name or Material 1-[2-Nitro-4-(trifluoromethyl)phenyl]piperazine
CAS 58315-38-1
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H12F3N3O2
Formula Weight 275.23
MDL Number MFCD00052613
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents