missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Naphthyl)ethanol, TCI America™

$132.92 - $430.84

Chemical Identifiers

CAS 7228-47-9
Molecular Formula C12H12O
Molecular Weight (g/mol) 172.227
MDL Number MFCD00004111
InChI Key AXRKCRWZRKETCK-UHFFFAOYSA-N
Synonym 1-2-naphthyl ethanol, 1-naphthalen-2-yl ethanol, +/--1-2-naphthyl ethanol, 1-naphthalen-2-yl ethan-1-ol, alpha-methylnaphthalene-2-methanol, .alpha.-methyl-2-naphthalenemethanol, pubchem9427, acmc-209gxz, acmc-1azul, methyl 2-naphtylcarbinol
PubChem CID 98243
IUPAC Name 1-naphthalen-2-ylethanol
SMILES CC(C1=CC2=CC=CC=C2C=C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N078825G View Documents TCI America
N078825G
25g Each for $430.84
 
N07885G View Documents TCI America
N07885G
5g Each for $132.92
 
Specifications

Chemical Identifiers

7228-47-9
172.227
AXRKCRWZRKETCK-UHFFFAOYSA-N
98243
CC(C1=CC2=CC=CC=C2C=C1)O
C12H12O
MFCD00004111
1-2-naphthyl ethanol, 1-naphthalen-2-yl ethanol, +/--1-2-naphthyl ethanol, 1-naphthalen-2-yl ethan-1-ol, alpha-methylnaphthalene-2-methanol, .alpha.-methyl-2-naphthalenemethanol, pubchem9427, acmc-209gxz, acmc-1azul, methyl 2-naphtylcarbinol
1-naphthalen-2-ylethanol

Specifications

170°C
76°C
7228-47-9
MFCD00004111
AXRKCRWZRKETCK-UHFFFAOYSA-N
1-naphthalen-2-ylethanol
98243
Crystal-Powder at 20°C
1-(2-Naphthyl)ethanol
White-Yellow
25g
C12H12O
1-2-naphthyl ethanol, 1-naphthalen-2-yl ethanol, +/--1-2-naphthyl ethanol, 1-naphthalen-2-yl ethan-1-ol, alpha-methylnaphthalene-2-methanol, .alpha.-methyl-2-naphthalenemethanol, pubchem9427, acmc-209gxz, acmc-1azul, methyl 2-naphtylcarbinol
CC(C1=CC2=CC=CC=C2C=C1)O
172.227
172.23
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No