Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Naphthyl)ethanol, 97%

$189.85 - $189.85

Chemical Identifiers

Chemical Name or Material 1-(2-Naphthyl)ethanol
CAS 7228-47-9
Molecular Formula C12H12O
Formula Weight 172.23
MDL Number MFCD00004111
Beilstein 1907449
Synonym alpha-Methyl-2-naphthalenemethanol; Methyl 2-naphthyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0510809

  SDS
alfa aesar™
L0510809
10g Each for $189.85
Description & Specifications

Specifications

Formula Weight 172.23
CAS 7228-47-9
Assay 97%
Chemical Name or Material 1-(2-Naphthyl)ethanol
Synonym alpha-Methyl-2-naphthalenemethanol; Methyl 2-naphthyl carbinol
Beilstein 1907449
Quantity 10g
Molecular Formula C12H12O
Boiling Point 170°C (13mmHg)
Melting Point 72° to 75°C
MDL Number MFCD00004111