missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Naphthoquinone, 95%, Tech., ACROS Organics™

Manufacturer:  Acros Organics 171230250

 View more versions of this product

Catalog No. AC171230250


 

Specifications

Specifications

1, 2-Naphthoquinone
136.0°C to 141.0°C
tech., 97%
94.0
Authentic
Technical Grade
95%
MFCD00001698
01,713
1, 2-Naphthalenedione
C1=CC=C2C(=C1)C=CC(=O)C2=O
158.156
CHEBI:34055
Crystalline Mass or Waxy Solid
Colorless or White to Yellow
25g
524-42-5
100.0
Glass bottle
95%
C10H6O2
07, 709
15, 6479
KETQAJRQOHHATG-UHFFFAOYSA-N
naphthalene-1,2-dione
10667
158.16

Chemical Identifiers

524-42-5
158.156
KETQAJRQOHHATG-UHFFFAOYSA-N
10667
C10H6O2
MFCD00001698
1, 2-Naphthalenedione
CHEBI:34055
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 208-360-2

RTECSNumber : QL7000000

TSCA : TSCA

SDS Documents