Learn More
1,2-Naphthoquinone-4-sulfonic acid sodium salt, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 415400250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1, 2-Naphthoquinone-4-sulfonic acid sodium salt | |
521-24-4 | |
100.0 | |
Yellow | |
97.5% min. (Iodometry) | |
C10H5NaO5S | |
25 g | |
15, 8777 | |
Solubility in water: 33.3g/L (20°C) | |
C1=CC=C2C(=C1)C(=CC(=O)C2=O)S(=O)(=O)[O-].[Na+] | |
260.19 | |
260.19 | |
Powder |
99% | |
98.5 | |
289.0°C | |
Authentic | |
Glass bottle | |
MFCD00001700 | |
02, 330 | |
1,2-naphthoquinone-4-sulfonic acid sodium salt, sodium 1,2-naphthoquinone-4-sulfonate, sodium beta-naphthoquinone-4-sulfonate, sodium 3,4-dioxonaphthalene-1-sulfonate, sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulfonate, unii-t7185hp5sh, sodium 3,4-dioxonaphthalene-1-sulphonate, beta-naphthoquinone-4-sulfonate sodium salt, 1-naphthalenesulfonic acid, 3,4-dihydro-3,4-dioxo-, sodium salt, chembl61245 | |
UBLXEEBHYISRFM-UHFFFAOYSA-M | |
sodium;3,4-dioxonaphthalene-1-sulfonate | |
516996 | |
98% |
Chemical Identifiers
521-24-4 | |
260.19 | |
UBLXEEBHYISRFM-UHFFFAOYSA-M | |
516996 | |
C1=CC=C2C(=C1)C(=CC(=O)C2=O)S(=O)(=O)[O-].[Na+] |
C10H5NaO5S | |
MFCD00001700 | |
1,2-naphthoquinone-4-sulfonic acid sodium salt, sodium 1,2-naphthoquinone-4-sulfonate, sodium beta-naphthoquinone-4-sulfonate, sodium 3,4-dioxonaphthalene-1-sulfonate, sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulfonate, unii-t7185hp5sh, sodium 3,4-dioxonaphthalene-1-sulphonate, beta-naphthoquinone-4-sulfonate sodium salt, 1-naphthalenesulfonic acid, 3,4-dihydro-3,4-dioxo-, sodium salt, chembl61245 | |
sodium;3,4-dioxonaphthalene-1-sulfonate |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 208-308-9
RTECSNumber : QK1490000
TSCA : TSCA
RUO – Research Use Only