missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1,2-Naphthalenedicarbonitrile, TCI America™

$268.89 - $895.30

Chemical Identifiers

CAS 19291-76-0
Molecular Formula C12H6N2
Molecular Weight (g/mol) 178.194
MDL Number MFCD00191673
InChI Key ZNPWYAMBOPRTHW-UHFFFAOYSA-N
Synonym 1,2-Dicyanonaphthalene
PubChem CID 350412
IUPAC Name naphthalene-1,2-dicarbonitrile
SMILES C1=CC=C2C(=C1)C=CC(=C2C#N)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N05781G View Documents TCI America
N05781G
1g Each for $268.89
 
N05785G View Documents TCI America
N05785G
5g Each for $895.30
 
Specifications

Chemical Identifiers

19291-76-0
178.194
ZNPWYAMBOPRTHW-UHFFFAOYSA-N
350412
C1=CC=C2C(=C1)C=CC(=C2C#N)C#N
C12H6N2
MFCD00191673
1,2-Dicyanonaphthalene
naphthalene-1,2-dicarbonitrile

Specifications

White-Yellow
1g
C12H6N2
3439
ZNPWYAMBOPRTHW-UHFFFAOYSA-N
naphthalene-1,2-dicarbonitrile
350412
Crystal-Powder at 20°C
1,2-Naphthalenedicarbonitrile
193°C
19291-76-0
MFCD00191673
1,2-Dicyanonaphthalene
C1=CC=C2C(=C1)C=CC(=C2C#N)C#N
178.194
178.19
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No