Please login to your online account to display your discounted pricing

1-(2-Methylphenyl)piperazine, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 1-(2-Methylphenyl)piperazine
CAS 39512-51-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H16N2
Formula Weight 176.26
MDL Number MFCD00040729
Synonym N-(o-Tolyl)piperazine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC382130010

  SDS
thermo fisher scientific
AC382130010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Low Melting Mass, Crystals or Crystalline Powder
Formula Weight 176.26
Quantity 1g
Color Off-white to light yellow
Melting Point 47° to 51°C
CAS Max % 100.0
Assay Percent Range 98%
Chemical Name or Material 1-(2-Methylphenyl)piperazine
MDL Number MFCD00040729
CAS Min % 97.5
Packaging Glass bottle
Synonym N-(o-Tolyl)piperazine
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
CAS 39512-51-1
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Molecular Formula C11H16N2
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.