Please login to your online account to display your discounted pricing

Alfa Aesar™ 1,2-Methylenedioxy-4-nitrobenzene, 98+%

$71.72 - $726.26

Chemical Identifiers

Chemical Name or Material 1,2-Methylenedioxy-4-nitrobenzene
CAS 2620-44-2
Molecular Formula C7H5NO4
Formula Weight 167.12
MDL Number MFCD00005824
Beilstein 169063
Synonym 5-Nitro-1,3-benzodioxole; 4-Nitro-1,2-(methylenedioxy)benzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2483806

  SDS
alfa aesar™
B2483806
5g Each for $71.72

AAB2483814

  SDS
alfa aesar™
B2483814
25g Each for $218.49

AAB2483822

  SDS
alfa aesar™
B2483822
100g Each for $726.26
Description & Specifications

Specifications

Formula Weight 167.12
Chemical Name or Material 1,2-Methylenedioxy-4-nitrobenzene
TSCA TSCA
Assay ≥98%
Molecular Formula C7H5NO4
MDL Number MFCD00005824
CAS 2620-44-2
Beilstein 169063
Synonym 5-Nitro-1,3-benzodioxole; 4-Nitro-1,2-(methylenedioxy)benzene
Health Hazard 1 Danger
Melting Point 142° to 149°C
Quantity 5g
Flash Point 142°C (287°F)
Health Hazard 2 GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.