Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-[(2-Methyl-4-thiazolyl)methyl]piperazine hydrochloride

$119.45 - $412.53

Chemical Identifiers

Chemical Name or Material 1-[(2-Methyl-4-thiazolyl)methyl]piperazine hydrochloride
CAS 880361-73-9
Molecular Formula C9H15N3S·HCl
Formula Weight 233.76
MDL Number MFCD07771967
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5102103

 
alfa aesar™
H5102103
1g Each for $412.53

AAH51021MD

 
alfa aesar™
H51021MD
250mg Each for $119.45
Description & Specifications

Specifications

Formula Weight 233.76
Chemical Name or Material 1-[(2-Methyl-4-thiazolyl)methyl]piperazine hydrochloride
Molecular Formula C9H15N3S·HCl
MDL Number MFCD07771967
Health Hazard 1 Warning
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 880361-73-9
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.