Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Methoxyphenyl)piperazine hydrochloride, 97%

Chemical Identifiers

Chemical Name or Material 1-(2-Methoxyphenyl)piperazine hydrochloride
CAS 5464-78-8
Molecular Formula C11H16N2O·HCl
Formula Weight 228.72
MDL Number MFCD00012762
UN Number UN2811
Beilstein 4276925
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2120514

 
alfa aesar™
B2120514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2120522

 
alfa aesar™
B2120522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2120536

 
alfa aesar™
B2120536
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 228.72
Chemical Name or Material 1-(2-Methoxyphenyl)piperazine hydrochloride
Assay 97%
Molecular Formula C11H16N2O·HCl
MDL Number MFCD00012762
CAS 5464-78-8
UN Number UN2811
Beilstein 4276925
Melting Point 213° to 216°C
Quantity 25g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Applications
1-(2-Methoxyphenyl)piperazine hydrochloride is used as reference material for forensic laboratories. It is used in chemical synthesis studies.

Solubility
Soluble in DMSO and Methanol.

Notes
Hygroscopic. Store at -20°C. Store away from oxidizing agents. Store in a cool, dry condition in a well sealed container.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.