Please login to your online account to display your discounted pricing

1-(2-Methoxyphenyl)piperazine 98+%, ACROS Organics™

$171.82 - $171.82

Chemical Identifiers

Chemical Name or Material 1-(2-Methoxyphenyl)piperazine
Name Note 98+%
CAS 35386-24-4
CAS Min % 98.0
CAS Max % 100.0
Assay 98.0 to 102.0%, 1.0% max. (o-anisidine) (GC)
Assay Percent Range 98+%
Molecular Formula C11H16N2O
Formula Weight 192.26
MDL Number MFCD00005958
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC126040250

  SDS
thermo fisher scientific
126040250
Glass bottle 25g Each for $171.82

AC126040050

  SDS
fisher scientific acros
126040050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC126041000

  SDS
thermo fisher scientific
AC126041000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid After Melting
Formula Weight 192.26
Quantity 25g
Color Clear colorless to yellow
EINECS Number 252-537-7
TSCA TSCA
Assay 98.0 to 102.0%, 1.0% max. (o-anisidine) (GC)
Density 1.09g/mL
CAS Min % 98.0
CAS Max % 100.0
Refractive Index 1.5745 to 1.5765 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
Packaging Glass bottle
Chemical Name or Material 1-(2-Methoxyphenyl)piperazine
Infrared Spectrum Authentic
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C11H16N2O
MDL Number MFCD00005958
CAS 35386-24-4
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.