Please login to your online account to display your discounted pricing

Alfa Aesar™ 1-(2-Methoxyphenyl)piperazine, 98%

Chemical Identifiers

Chemical Name or Material 1-(2-Methoxyphenyl)piperazine
CAS 35386-24-4
Molecular Formula C11H16N2O
Formula Weight 192.26
MDL Number MFCD00005958
UN Number UN2923
Beilstein 167888
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1581914

  SDS
alfa aesar™
A1581914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1581922

  SDS
alfa aesar™
A1581922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 192.26
MDL Number MFCD00005958
Beilstein 167888
Flash Point >110°C (230°F)
Assay 98%
Molecular Formula C11H16N2O
CAS 35386-24-4
Melting Point 35° to 39°C
Chemical Name or Material 1-(2-Methoxyphenyl)piperazine
UN Number UN2923
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Boiling Point 130° to 133°C (0.1mmHg)
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Quantity 25g

Applications
It is used as pharmaceutical intermediate. N-alkylated 1-(2-methyoxyphenyl)piperazines markedly improved affinity and selectivity of the dopamine D3 receptor which is recognized as a potential therapeutic target for the treatment of various neurological and psychiatric disorders.

Solubility
Soluble in chloroform, ethyl acetate, and methanol. Insoluble in water

Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.