missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Methoxyphenoxy)-2-propanol, TCI America™

Manufacturer:  TCI America M10605G

 View more versions of this product

Catalog No. M10605G



Specifications

Specifications

1-(2-Methoxyphenoxy)-2-propanol
5g
C10H14O3
2-(2-Hydroxypropoxy)anisole
CC(COC1=CC=CC=C1OC)O
182.219
182.22
≥90.0% (GC)
Colorless
64120-49-6
MFCD00191539
CUJJTCXPBXHBTJ-UHFFFAOYSA-N
1-(2-methoxyphenoxy)propan-2-ol
22036327
Clear Liquid at 20°C

Chemical Identifiers

64120-49-6
182.219
CUJJTCXPBXHBTJ-UHFFFAOYSA-N
22036327
CC(COC1=CC=CC=C1OC)O
C10H14O3
MFCD00191539
2-(2-Hydroxypropoxy)anisole
1-(2-methoxyphenoxy)propan-2-ol
Safety and Handling

Safety and Handling

TSCA : No

Documents