missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

1-(2-Methoxyethyl)piperazine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC367110010

 View more versions of this product

Catalog No. AC367110010



Description

Description

Specifications

Specifications

1-(2-Methoxyethyl)piperazine
87°C
Glass bottle
0.97
Colorless to Yellow
98%
97.5
97.5% min. (GC)
MFCD00191214
BMEMBBFDTYHTLH-UHFFFAOYSA-N
1-(2-methoxyethyl)piperazine
2734638
Liquid
0.9700g/mL
Authentic
1.4730 to 1.475
193°C to 194°C
1g
13484-40-7
100.0
C7H16N2O
1-2-methoxy-ethyl-piperazine, 1-2-methoxyethyl piperazine, 1-2-methoxyethyl-piperazine, 1-methoxy-2-piperazinylethane, 2-methoxyethyl piperazine, 2-methoxyethylpiperazine, n-methoxyethyl piperazine, n-methoxyethylylpiperazine, piperazine, 1-2-methoxyethyl, pubchem14209
COCCN1CCNCC1
144.21
144.21
98%

Chemical Identifiers

13484-40-7
144.21
BMEMBBFDTYHTLH-UHFFFAOYSA-N
2734638
COCCN1CCNCC1
C7H16N2O
MFCD00191214
1-2-methoxy-ethyl-piperazine, 1-2-methoxyethyl piperazine, 1-2-methoxyethyl-piperazine, 1-methoxy-2-piperazinylethane, 2-methoxyethyl piperazine, 2-methoxyethylpiperazine, n-methoxyethyl piperazine, n-methoxyethylylpiperazine, piperazine, 1-2-methoxyethyl, pubchem14209
1-(2-methoxyethyl)piperazine
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

SDS Documents